Assay Details
Target Gene Details
Entrez Gene ID: | 18104 |
Gene Name: | NAD(P)H dehydrogenase, quinone 1 |
Gene Aliases: |
AV001255, Dia4, Dtd, Nmo-1, Nmo1, Nmor1, Ox-1, Ox1, Qr1 |
Location: |
Chr.8:107388225-107403205 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nqo1 | NM_008706.5 | NP_032732.3 | ||
| AK075568.1 | BAC35826.1 | |||
| BC004579.1 | AAH04579.1 | |||
| CT010284.1 | CAJ18492.1 | |||
| S75951.1 | AAB32718.1 | |||
| U12961.1 | AAA80184.1 |
More Information
Set Membership: |
|

Genomic Map