Assay Details
Target Gene Details
Entrez Gene ID: | 210801 |
Gene Name: | unc-5 homolog D (C. elegans) |
Gene Aliases: |
D930029E11Rik, Unc5h4, mKIAA1777 |
Location: |
Chr.8:28646717-29219682 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc5d | NM_153135.3 | 14 | 2315 | NP_694775.1 |
| AJ487854.1 | 14 | 2316 | CAD32252.1 | |
| AK220417.1 | 4 | 656 | BAD90463.1 | |
| BC137615.1 | 14 | 2106 | ||
| BC144783.1 | 12 | 1923 | ||
| BC144784.1 | 12 | 1884 | ||
| BC145695.1 | 11 | 1890 | ||
| FJ824810.1 | 13 | 2194 |
More Information
Set Membership: |
|

Genomic Map