Assay Details
Target Gene Details
Entrez Gene ID: | 210801 |
Gene Name: | unc-5 homolog D (C. elegans) |
Gene Aliases: |
D930029E11Rik, Unc5h4, mKIAA1777 |
Location: |
Chr.8:28646717-29219682 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Unc5d | NM_153135.3 | NP_694775.1 | ||
| AJ487854.1 | CAD32252.1 | |||
| BC137615.1 | ||||
| BC144783.1 | ||||
| BC144784.1 | ||||
| BC145695.1 | ||||
| FJ824810.1 |
More Information
Set Membership: |
|

Genomic Map