Assay Details
Target Gene Details
Entrez Gene ID: | 94187 |
Gene Name: | zinc finger protein 423 |
Gene Aliases: |
Ebfaz, Roaz, Zfp104, Znf423, ataxia1, mKIAA0760, nur12 |
Location: |
Chr.8:87661810-87961859 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp423 | NM_033327.2 | 3 | 609 | NP_201584.2 |
| AF188609.1 | AAG17053.1 | |||
| AK122365.1 | 3 | 575 | BAC65647.1 | |
| AY147407.1 | 2 | 214 | AAN39840.1 | |
| AY256893.1 | 3 | 508 | AAP33073.1 | |
| BC059234.1 | 1 | 420 | AAH59234.1 | |
| BC139028.1 | 3 | 355 | ||
| BC139030.1 | 3 | 355 | ||
| BQ180174.1 | ||||
| CL210542.2 | 3 | 260 |
More Information
Set Membership: |
|

Genomic Map