Assay Details
Target Gene Details
Entrez Gene ID: | 13070 |
Gene Name: | cytochrome P450, family 11, subfamily a, polypeptide 1 |
Gene Aliases: |
Cyp11a, Cypxia1, D9Ertd411e, P450scc, Scc, cscc |
Location: |
Chr.9:58015017-58027023 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp11a1 | NM_019779.3 | 4 | 776 | NP_062753.3 |
AF195119.1 | 4 | 776 | AAF03897.1 | |
AK077945.1 | 3 | 1696 | ||
BC068264.1 | 4 | 795 | AAH68264.1 |
