Assay Details
Target Gene Details
Entrez Gene ID: | 66595 |
Gene Name: | asteroid homolog 1 (Drosophila) |
Gene Aliases: |
1100001A21Rik, AV006403 |
Location: |
Chr.9:105395294-105408298 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aste1 | NM_025651.4 | 1 | 154 | NP_079927.2 |
| AK030661.1 | BAC27067.1 | |||
| BC098471.1 | 1 | 131 | AAH98471.1 |
Target Gene Details
Entrez Gene ID: | 208583 |
Gene Name: | NIMA (never in mitosis gene a)-related expressed kinase 11 |
Gene Aliases: |
4932416N14Rik |
Location: |
Chr.9:105162156-105395524 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nek11 | AK043543.1 | BAC31576.1 |
More Information
Set Membership: |
|

Genomic Map