Assay Details
Target Gene Details
Entrez Gene ID: | 66595 |
Gene Name: | asteroid homolog 1 (Drosophila) |
Gene Aliases: |
1100001A21Rik, AV006403 |
Location: |
Chr.9:105395294-105408298 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aste1 | NM_001164828.1 | 3 | 380 | NP_001158300.1 |
NM_025651.4 | 4 | 1897 | NP_079927.2 | |
AK003146.1 | 3 | 1234 | BAB22602.1 | |
AK006153.1 | 3 | 382 | ||
AK030661.1 | 4 | 1768 | BAC27067.1 | |
BC048866.1 | 3 | 358 | AAH48866.1 | |
BC098471.1 | 5 | 1782 | AAH98471.1 |
