Assay Details
Target Gene Details
Entrez Gene ID: | 110446 |
Gene Name: | acetyl-Coenzyme A acetyltransferase 1 |
Gene Aliases: |
6330585C21Rik, Acat |
Location: |
Chr.9:53580522-53610350 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acat1 | NM_144784.3 | NP_659033.1 | ||
| AJ510150.1 | CAD52869.1 | |||
| AK032097.1 | BAC27697.1 | |||
| AK075593.1 | ||||
| AK081715.1 | BAC38304.1 | |||
| AK146241.1 | ||||
| AK163402.1 | BAE37335.1 | |||
| AK169771.1 | BAE41356.1 | |||
| BC024763.1 | AAH24763.1 |
More Information
Set Membership: |
|

Genomic Map