Assay Details
Target Gene Details
Entrez Gene ID: | 67213 |
Gene Name: | CKLF-like MARVEL transmembrane domain containing 6 |
Gene Aliases: |
2810051A14Rik, AA536733, Cklfsf6 |
Location: |
Chr.9:114731157-114749347 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cmtm6 | NM_026036.3 | 3 | 395 | NP_080312.1 |
| AK012941.1 | 3 | 396 | BAB28562.1 | |
| AK030885.1 | 3 | 375 | BAC27172.1 | |
| AK128960.1 | 3 | 381 | ||
| AK145743.1 | 3 | 376 | BAE26621.1 | |
| AK150775.1 | 3 | 396 | ||
| AK151092.1 | 3 | 396 | ||
| AK151431.1 | 3 | 396 | ||
| AK152448.1 | 3 | 397 | BAE31227.1 | |
| AK152723.1 | 3 | 406 | BAE31446.1 | |
| AK152817.1 | 3 | 406 | BAE31520.1 | |
| AK153092.1 | 3 | 396 | ||
| AK154481.1 | 3 | 382 | BAE32617.1 | |
| AK159439.1 | 3 | 365 | BAE35085.1 | |
| AK159735.1 | 3 | 390 | BAE35329.1 | |
| AK165948.1 | 3 | 364 | BAE38480.1 | |
| AK167499.1 | 3 | 382 | BAE39575.1 | |
| AK167854.1 | 3 | 387 | BAE39872.1 | |
| AK170905.1 | 3 | 397 | BAE42106.1 | |
| AK171394.1 | 3 | 426 | BAE42431.1 | |
| AY241868.1 | 3 | 432 | AAP33490.1 | |
| BC007194.1 | AAH07194.1 | |||
| BC027248.1 | 3 | 367 | AAH27248.1 |
More Information
Set Membership: |
|

Genomic Map