Assay Details
Target Gene Details
Entrez Gene ID: | 100529079 |
Gene Name: | RIKEN cDNA 5830418P13 gene |
Gene Aliases: |
- |
Location: |
Chr.9:103423578-103461260 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5830418P13Rik | NR_040466.1 | |||
| AK079871.1 | ||||
| AK134047.1 |
Target Gene Details
Entrez Gene ID: | 107993 |
Gene Name: | beaded filament structural protein 2, phakinin |
Gene Aliases: |
AI448593, CP49 |
Location: |
Chr.9:103424924-103480414 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bfsp2 | NM_001002896.2 | NP_001002896.1 | ||
| AK134810.1 | BAE22294.1 | |||
| AK198043.1 | ||||
| BC068172.1 | AAH68172.1 |
More Information
Set Membership: |
|

Genomic Map