Assay Details
Target Gene Details
Entrez Gene ID: | 100529079 |
Gene Name: | RIKEN cDNA 5830418P13 gene |
Gene Aliases: |
- |
Location: |
Chr.9:103423578-103461260 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5830418P13Rik | NR_040466.1 | 2 | 182 | |
| AK079871.1 | ||||
| AK134047.1 | 2 | 183 |
Target Gene Details
Entrez Gene ID: | 107993 |
Gene Name: | beaded filament structural protein 2, phakinin |
Gene Aliases: |
AI448593, CP49 |
Location: |
Chr.9:103424924-103480414 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bfsp2 | NM_001002896.2 | 6 | 1200 | NP_001002896.1 |
| AK134810.1 | 6 | 1132 | BAE22294.1 | |
| AK178846.1 | ||||
| AK195636.1 | ||||
| BC062815.1 | 4 | 402 | AAH62815.1 | |
| BC068172.1 | 6 | 1200 | AAH68172.1 | |
| S55549.1 | 4 | 509 | AAB25419.1 |

Genomic Map