Assay Details
Target Gene Details
Entrez Gene ID: | 13663 |
Gene Name: | etoposide induced 2.4 mRNA |
Gene Aliases: |
AA536736, AI115355, PIG8 |
Location: |
Chr.9:36779153-36797393 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ei24 | NM_001199494.1 | NP_001186423.1 | ||
NM_007915.5 | NP_031941.1 | |||
AK145018.1 | ||||
AK147776.1 | BAE28130.1 | |||
AK159927.1 | BAE35489.1 | |||
AK172233.1 | BAE42899.1 | |||
U41751.2 | AAC52483.2 |
