Assay Details
Target Gene Details
Entrez Gene ID: | 68969 |
Gene Name: | eukaryotic translation initiation factor 1B |
Gene Aliases: |
1500010M16Rik, 3110001N14Rik, Gc20 |
Location: |
Chr.9:120492606-120495327 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif1b | NM_026892.3 | NP_081168.1 | ||
| AI154212.1 | ||||
| AK005192.1 | BAB23874.1 | |||
| AK013965.1 | BAB29089.1 | |||
| BC030319.1 | AAH30319.1 | |||
| BC033505.1 | AAH33505.1 |
More Information
Set Membership: |
|

Genomic Map