Assay Details
Target Gene Details
Entrez Gene ID: | 113868 |
Gene Name: | acetyl-Coenzyme A acyltransferase 1A |
Gene Aliases: |
Acaa, Acaa1, D9Ertd25e, PTL |
Location: |
Chr.9:119341052-119350296 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acaa1a | NM_130864.3 | NP_570934.1 | ||
| AK143187.1 | BAE25296.1 | |||
| AY273811.1 | AAP31668.1 | |||
| BC012400.1 | AAH12400.1 |
More Information
Set Membership: |
|

Genomic Map