Assay Details
Target Gene Details
Entrez Gene ID: | 235674 |
Gene Name: | acetyl-Coenzyme A acyltransferase 1B |
Gene Aliases: |
- |
Location: |
Chr.9:119148043-119157093 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acaa1b | NM_146230.3 | NP_666342.1 | ||
| AK145953.1 | BAE26781.1 | |||
| AK160568.1 | BAE35878.1 | |||
| AY273812.1 | AAP31669.1 | |||
| BC019882.1 | AAH19882.1 |
More Information
Set Membership: |
|

Genomic Map