Assay Details
Target Gene Details
Entrez Gene ID: | 69524 |
Gene Name: | endothelial cell-specific adhesion molecule |
Gene Aliases: |
2310008D05Rik, Esam1, W117m |
Location: |
Chr.9:37528055-37538319 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Esam | NM_027102.3 | NP_081378.1 | ||
AF361882.1 | AAK51504.1 | |||
AK009223.1 | ||||
AK156399.1 | BAE33699.1 | |||
BC019460.1 | ||||
BC049249.1 | AAH49249.1 | |||
BQ043033.1 | ||||
CJ048383.1 |
