Assay Details
Target Gene Details
Entrez Gene ID: | 69524 |
Gene Name: | endothelial cell-specific adhesion molecule |
Gene Aliases: |
2310008D05Rik, Esam1, W117m |
Location: |
Chr.9:37528055-37538319 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Esam | NM_027102.3 | 6 | 929 | NP_081378.1 |
AF361882.1 | 6 | 914 | AAK51504.1 | |
AK009223.1 | 6 | 904 | ||
AK156399.1 | 6 | 927 | BAE33699.1 | |
BC019460.1 | 6 | 924 | ||
BC049249.1 | 6 | 909 | AAH49249.1 | |
BG077536.2 | 4 | 363 |
