Assay Details
Target Gene Details
Entrez Gene ID: | 74498 |
Gene Name: | golgi reassembly stacking protein 1 |
Gene Aliases: |
5430411C10Rik, GOLPH5, GRASP65, P65 |
Location: |
Chr.9:119925673-119937558 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gorasp1 | NM_028976.2 | 7 | 852 | NP_083252.1 |
AK154986.1 | 7 | 829 | BAE32975.1 | |
AK170627.1 | 7 | 816 | BAE41921.1 | |
AK196847.1 | 2 | 193 | ||
BC012251.1 | 7 | 852 | AAH12251.1 |
