Assay Details
Target Gene Details
Entrez Gene ID: | 68265 |
Gene Name: | IQ motif containing F3 |
Gene Aliases: |
1700012F17Rik, AV040440 |
Location: |
Chr.9:106504666-106561631 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqcf3 | NM_026645.3 | NP_080921.2 | ||
| AK016688.1 | BAB30380.1 | |||
| AK018835.1 | ||||
| AK132739.1 | BAE21327.1 | |||
| BC049549.1 | AAH49549.1 | |||
| DQ153247.1 | AAZ78215.1 |

Genomic Map