Assay Details
Target Gene Details
Entrez Gene ID: | 104360 |
Gene Name: | insulin related protein 2 (islet 2) |
Gene Aliases: |
3110001N10Rik |
Location: |
Chr.9:55538677-55547667 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Isl2 | NM_027397.3 | 2 | 288 | NP_081673.2 |
| AK013964.1 | 2 | 231 | BAB29088.1 | |
| BC120880.2 | 2 | 231 | ||
| BC148307.2 | 2 | 231 |

Genomic Map