Assay Details
Target Gene Details
Entrez Gene ID: | 26968 |
Gene Name: | immunoglobulin superfamily containing leucine-rich repeat |
Gene Aliases: |
AW553146 |
Location: |
Chr.9:58156246-58159239 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Islr | NM_012043.4 | NP_036173.1 | ||
BC006602.1 | AAH06602.1 |
