Assay Details
Target Gene Details
Entrez Gene ID: | 215194 |
Gene Name: | KRI1 homolog (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.9:21273457-21287969 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kri1 | NM_145416.3 | NP_663391.2 | ||
AK030447.1 | 1 | 292 | ||
BC021438.1 | AAH21438.1 | |||
BI989088.1 |
