Assay Details
Target Gene Details
Entrez Gene ID: | 72774 |
Gene Name: | nei endonuclease VIII-like 1 (E. coli) |
Gene Aliases: |
2810450N13Rik, Nei1 |
Location: |
Chr.9:57142788-57148675 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neil1 | NM_028347.2 | NP_082623.1 | ||
| AB079069.1 | BAC06477.1 | |||
| AK013322.1 | BAB28790.1 | |||
| AK155405.1 | BAE33246.1 | |||
| AK219577.1 | ||||
| BC043297.1 | AAH43297.1 |
More Information
Set Membership: |
|

Genomic Map