Assay Details
Target Gene Details
Entrez Gene ID: | 16173 |
Gene Name: | interleukin 18 |
Gene Aliases: |
Igif, Il-18 |
Location: |
Chr.9:50554705-50581841 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il18 | NM_008360.1 | NP_032386.1 | ||
| AK078100.1 | ||||
| AY157834.1 | AAO21309.1 | |||
| AY157835.1 | AAO21310.1 | |||
| BC024384.1 | AAH24384.1 | |||
| D49949.1 | BAA08705.1 |
More Information
Set Membership: |
|

Genomic Map