Assay Details
Target Gene Details
Entrez Gene ID: | 80907 |
Gene Name: | lactamase, beta |
Gene Aliases: |
LACT-1, Lact1, Mrpl56 |
Location: |
Chr.9:66955393-66975484 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lactb | NM_030717.1 | NP_109642.1 | ||
AF317900.1 | AAG37910.1 | |||
AK078550.1 | ||||
BC046293.1 | AAH46293.1 | |||
BC147809.1 |
