Assay Details
Target Gene Details
Entrez Gene ID: | 28015 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide M |
Gene Aliases: |
AA407243, AA408799, AI303746, AW553238, D9Wsu138e, Grinl1a, mKIAA4185 |
Location: |
Chr.9:71478437-71485983 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polr2m | NM_001164793.1 | 3 | 933 | NP_001158265.1 |
NM_178602.3 | 3 | 1009 | NP_848717.1 | |
AK002571.1 | 3 | 934 | BAB22196.1 | |
AK003433.1 | ||||
AK014316.1 | 3 | 941 | BAB29267.1 | |
AK017773.1 | 3 | 952 | ||
AK051868.1 | 3 | 943 | BAC34793.1 | |
AK052037.1 | 3 | 963 | BAC34836.1 | |
AK078750.1 | 3 | 944 | ||
AK079119.1 | 3 | 927 | BAC37548.1 | |
AK136806.1 | 3 | 961 | ||
AK151221.1 | 3 | 947 | ||
AK159924.1 | 3 | 956 | BAE35486.1 | |
AK166738.1 | 3 | 956 | BAE38981.1 | |
BC018502.1 | 3 | 935 | AAH18502.1 | |
BC062199.1 | 3 | 929 | AAH62199.1 |
