Assay Details
Target Gene Details
Entrez Gene ID: | 67557 |
Gene Name: | La ribonucleoprotein domain family, member 6 |
Gene Aliases: |
5430431G03Rik, AI552438, Achn |
Location: |
Chr.9:60713121-60738801 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Larp6 | NM_026235.4 | NP_080511.2 | ||
| AK017372.1 | BAB30713.1 | |||
| AK020966.1 | BAB32263.1 | |||
| AK039614.1 | BAC30400.1 | |||
| AK042616.1 | BAC31306.1 | |||
| AK087521.1 | BAC39909.1 | |||
| BC090615.1 | AAH90615.1 |
More Information
Set Membership: |
|

Genomic Map