Assay Details
Target Gene Details
Entrez Gene ID: | 110119 |
Gene Name: | mannose phosphate isomerase |
Gene Aliases: |
1110002E17Rik, AI315153, Mpi-1, Mpi1 |
Location: |
Chr.9:57544260-57552812 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpi | NM_025837.2 | NP_080113.1 | ||
| AF244360.1 | AAK69388.1 | |||
| AK003289.1 | ||||
| AK003896.1 | ||||
| AK132777.1 | BAE21353.1 | |||
| BC009068.1 | AAH09068.1 | |||
| BC051163.1 | AAH51163.1 |
More Information
Set Membership: |
|

Genomic Map