Assay Details
Target Gene Details
Entrez Gene ID: | 110119 |
Gene Name: | mannose phosphate isomerase |
Gene Aliases: |
1110002E17Rik, AI315153, Mpi-1, Mpi1 |
Location: |
Chr.9:57544260-57552812 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpi | NM_025837.2 | 7 | 946 | NP_080113.1 |
| AF244360.1 | 7 | 928 | AAK69388.1 | |
| AK003289.1 | 5 | 706 | ||
| AK003896.1 | 7 | 947 | ||
| AK132777.1 | 7 | 947 | BAE21353.1 | |
| AK189289.1 | 2 | 95 | ||
| BC009068.1 | 7 | 924 | AAH09068.1 | |
| BC051163.1 | 7 | 924 | AAH51163.1 |
More Information
Set Membership: |
|

Genomic Map