Assay Details
Target Gene Details
Entrez Gene ID: | 208098 |
Gene Name: | pannexin 3 |
Gene Aliases: |
3230401P04, 4833413G11Rik |
Location: |
Chr.9:37659902-37669222 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Panx3 | NM_172454.2 | NP_766042.2 | ||
AK028288.1 | BAC25860.1 | |||
AK036933.1 | BAC29644.1 | |||
BC052822.1 | AAH52822.1 |
