Assay Details
Target Gene Details
Entrez Gene ID: | 57014 |
Gene Name: | 5-hydroxytryptamine (serotonin) receptor 3B |
Gene Aliases: |
5-HT3B |
Location: |
Chr.9:48935008-48964990 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Htr3b | NM_020274.4 | 8 | 1197 | NP_064670.1 |
| AF155045.1 | 8 | 1029 | AAF73284.1 | |
| AK085213.1 | 8 | 1198 | BAC39391.1 | |
| BC046990.1 | 8 | 1175 | AAH46990.1 |
More Information
Set Membership: |
|

Genomic Map