Assay Details
Target Gene Details
Entrez Gene ID: | 102680 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 20A |
Gene Aliases: |
A730081N20Rik, AU022428, Xt3s1, Xtrp3s1 |
Location: |
Chr.9:123636907-123678832 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a20a | NM_139142.2 | 8 | 1233 | NP_631881.1 |
| AJ964961.1 | 8 | 1207 | CAI80736.1 | |
| AK043297.1 | 8 | 1244 | ||
| BC118933.2 | 8 | 1233 |
More Information
Set Membership: |
|

Genomic Map