Assay Details
Target Gene Details
Entrez Gene ID: | 22273 |
Gene Name: | ubiquinol-cytochrome c reductase core protein 1 |
Gene Aliases: |
1110032G10Rik |
Location: |
Chr.9:108936648-108949641 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uqcrc1 | NM_025407.2 | NP_079683.2 | ||
| AI462428.1 | ||||
| AK002292.1 | ||||
| AK004031.1 | ||||
| AK010553.1 | BAB27022.1 | |||
| AK013128.1 | BAB28666.1 | |||
| AK151764.1 | BAE30670.1 | |||
| AK160337.1 | BAE35744.1 | |||
| AK167911.1 | BAE39918.1 | |||
| AK168218.1 | BAE40175.1 | |||
| AK178594.1 | ||||
| AK183917.1 | ||||
| AK189225.1 | ||||
| AK212551.1 | ||||
| AK214273.1 | ||||
| AK215619.1 | ||||
| BC018405.1 | AAH18405.1 | |||
| BC030064.1 | AAH30064.1 |
More Information
Set Membership: |
|

Genomic Map