Assay Details
Target Gene Details
Entrez Gene ID: | 20347 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B |
Gene Aliases: |
SemA, Semaa, sema5, semaV |
Location: |
Chr.9:107597674-107609303 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema3b | NM_001042779.2 | NP_001036244.1 | ||
| NM_001291537.1 | NP_001278466.1 | |||
| NM_001291538.1 | NP_001278467.1 | |||
| NM_001291539.1 | NP_001278468.1 | |||
| NM_009153.3 | NP_033179.2 | |||
| NR_111986.1 | ||||
| AK014333.1 | ||||
| AK033597.1 | ||||
| AK037034.1 | ||||
| AK043634.1 | ||||
| AK048364.1 | ||||
| AK148232.1 | ||||
| AK185992.1 | ||||
| AK199289.1 | ||||
| AK205569.1 | ||||
| BB116052.2 | ||||
| BC090669.1 | ||||
| BC150758.1 | ||||
| X85990.1 | CAA59982.1 |
More Information
Set Membership: |
|

Genomic Map