Assay Details
Target Gene Details
Entrez Gene ID: | 67629 |
Gene Name: | SPC24, NDC80 kinetochore complex component, homolog (S. cerevisiae) |
Gene Aliases: |
2410030K01Rik, AV109292, Spbc24 |
Location: |
Chr.9:21755442-21760452 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spc24 | NM_026282.5 | NP_080558.1 | ||
AK011728.1 | BAB27804.1 | |||
AK031680.1 | BAC27510.1 | |||
AK075950.1 | BAC36077.1 | |||
AK153185.1 | BAE31786.1 | |||
AK167877.1 | BAE39891.1 | |||
BC086683.1 | ||||
BC116238.1 | ||||
BC116239.1 |
