Assay Details
Target Gene Details
Entrez Gene ID: | 67049 |
Gene Name: | pseudouridine synthase 3 |
Gene Aliases: |
2610020J05Rik, 5730412F04Rik |
Location: |
Chr.9:35558597-35567400 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pus3 | AK030639.1 | 2 | 243 |
