Assay Details
Target Gene Details
Entrez Gene ID: | 80893 |
Gene Name: | transmembrane protease, serine 5 (spinesin) |
Gene Aliases: |
spinesin |
Location: |
Chr.9:49080439-49117591 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmprss5 | NM_030709.2 | NP_109634.2 | ||
| AB016229.1 | BAB20276.1 | |||
| AB016230.1 | BAB20277.1 | |||
| AB016423.1 | BAB20278.1 | |||
| AB041037.1 | BAB40328.1 | |||
| AK029714.1 | BAC26577.1 | |||
| BC132151.1 | ||||
| BC145027.1 |
More Information
Set Membership: |
|

Genomic Map