Assay Details
Target Gene Details
Entrez Gene ID: | 21376 |
Gene Name: | transforming growth factor beta regulated gene 1 |
Gene Aliases: |
AA408552, AA409675, Niam, Tb-5, mFLJ00213 |
Location: |
Chr.9:37649182-37657312 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbrg1 | NM_025289.3 | NP_079565.2 | ||
| AK003270.1 | ||||
| AK010925.1 | ||||
| AK011268.1 | ||||
| AK131158.1 | 4 | 2364 | BAD21408.1 | |
| AK150549.1 | BAE29651.1 | |||
| AK151075.1 | BAE30090.1 | |||
| AK151209.1 | BAE30204.1 | |||
| AK151457.1 | BAE30416.1 | |||
| AK166547.1 | BAE38845.1 | |||
| AK205215.1 | ||||
| BC021331.1 | AAH21331.1 | |||
| BC040813.1 | ||||
| BC043022.1 | ||||
| BC065795.1 | AAH65795.1 | |||
| BQ174234.1 | ||||
| DQ144541.1 | AAZ76015.1 |
More Information
Set Membership: |
|

Genomic Map