Assay Details
Target Gene Details
Entrez Gene ID: | 235033 |
Gene Name: | retinol dehydrogenase 8 |
Gene Aliases: |
Gm182, prRDH |
Location: |
Chr.9:20818504-20826163 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdh8 | NM_001030290.1 | 5 | 627 | NP_001025461.1 |
More Information
Set Membership: |
|

Genomic Map