Assay Details
Target Gene Details
Entrez Gene ID: | 19089 |
Gene Name: | protein kinase C substrate 80K-H |
Gene Aliases: |
80K-H, PKCSH |
Location: |
Chr.9:22002988-22014245 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkcsh | NM_001293650.1 | NP_001280579.1 | ||
| NM_001293651.1 | NP_001280580.1 | |||
| NM_008925.2 | NP_032951.1 | |||
| AK137661.1 | BAE23450.1 | |||
| AK153931.1 | BAE32262.1 | |||
| AK159705.1 | BAE35304.1 | |||
| BC009816.1 | AAH09816.1 | |||
| CD807078.1 | ||||
| U92794.1 | AAC53183.1 |
More Information
Set Membership: |
|

Genomic Map