Assay Details
Target Gene Details
Entrez Gene ID: | 27966 |
Gene Name: | RRP9, small subunit (SSU) processome component, homolog (yeast) |
Gene Aliases: |
55kDa, D19435, D9Wsu10e, Rnu3ip2, U3-55k |
Location: |
Chr.9:106477309-106485415 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rrp9 | NM_145620.4 | NP_663595.1 | ||
AF368232.1 | AAN62792.1 | |||
AK076113.1 | BAC36193.1 | |||
AK156199.1 | ||||
BC014703.1 | AAH14703.1 |
