Assay Details
Target Gene Details
Entrez Gene ID: | 60409 |
Gene Name: | trafficking protein particle complex 4 |
Gene Aliases: |
1500017G03Rik, AI303617, HSPC172, PTD009, Sbd, Sbdn, Sdcbp2, TRS23 |
Location: |
Chr.9:44403759-44407563 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc4 | NM_021789.2 | NP_068561.1 | ||
| AF233340.1 | AAG24950.1 | |||
| AK005276.1 | BAE43192.1 | |||
| AK032168.1 | BAC27737.1 | |||
| BC038898.1 | AAH38898.1 |
More Information
Set Membership: |
|

Genomic Map