Assay Details
Target Gene Details
Entrez Gene ID: | 244954 |
Gene Name: | protease, serine 35 |
Gene Aliases: |
6030424L22Rik, P3D9 |
Location: |
Chr.9:86743240-86758443 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss35 | NM_178738.3 | NP_848853.2 | ||
| AK030671.1 | BAC27073.1 | |||
| AK031411.1 | BAC27392.1 | |||
| AK031644.1 | BAC27491.1 | |||
| AK076281.1 | ||||
| BB659182.1 | ||||
| BC075675.1 | AAH75675.1 | |||
| DQ223037.1 |
More Information
Set Membership: |
|

Genomic Map