Assay Details
Target Gene Details
Entrez Gene ID: | 74766 |
Gene Name: | Yip1 domain family, member 2 |
Gene Aliases: |
1300010K09Rik |
Location: |
Chr.9:21588682-21592831 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Yipf2 | NM_001205157.1 | 3 | 213 | NP_001192086.1 |
| NM_001205158.1 | 3 | 203 | NP_001192087.1 | |
| NM_138303.2 | 2 | 351 | NP_612176.1 | |
| NR_038062.1 | 2 | 152 | ||
| AA967152.1 | 2 | 150 | ||
| AK004959.1 | 2 | 137 | ||
| AK033705.1 | 3 | 162 | BAC28437.1 | |
| AK133631.1 | 3 | 173 | BAE21759.1 | |
| BC002282.1 | 2 | 266 | AAH02282.1 | |
| BC003289.1 | 3 | 144 | AAH03289.1 |

Genomic Map