Assay Details
Target Gene Details
Entrez Gene ID: | 74766 |
Gene Name: | Yip1 domain family, member 2 |
Gene Aliases: |
1300010K09Rik |
Location: |
Chr.9:21588682-21592831 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Yipf2 | NM_001205157.1 | NP_001192086.1 | ||
| NM_001205158.1 | NP_001192087.1 | |||
| NM_138303.2 | NP_612176.1 | |||
| NR_038062.1 | ||||
| AA967152.1 | ||||
| AK004959.1 | ||||
| AK033705.1 | BAC28437.1 | |||
| AK133631.1 | BAE21759.1 | |||
| BC002282.1 | AAH02282.1 | |||
| BC003289.1 | AAH03289.1 |
More Information
Set Membership: |
|

Genomic Map