Assay Details
Target Gene Details
Entrez Gene ID: | 319586 |
Gene Name: | CUGBP, Elav-like family member 5 |
Gene Aliases: |
4930565A21Rik, Brunol5 |
Location: |
Chr.10:81459227-81482725 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Celf5 | NM_176954.3 | 8 | 961 | NP_795928.2 |
| AK029796.1 | 8 | 2135 | ||
| AK132321.1 | 8 | 927 | ||
| AK158669.1 | 7 | 738 |

Genomic Map