Assay Details
Target Gene Details
Entrez Gene ID: | 17684 |
Gene Name: | Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2 |
Gene Aliases: |
AI835299, ER154-like, Mrg1, Msg2, p35srj |
Location: |
Chr.10:17723205-17725674 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cited2 | NM_010828.3 | NP_034958.2 | ||
| AK131664.1 | BAE20751.1 | |||
| AK133791.1 | BAE21844.1 | |||
| AK183582.1 | ||||
| AK187598.1 | ||||
| AK196895.1 | ||||
| BC057126.1 | AAH57126.1 | |||
| U86445.1 | AAC39945.1 | |||
| Y15163.1 | CAA75432.1 |
More Information
Set Membership: |
|

Genomic Map