Assay Details
Target Gene Details
Entrez Gene ID: | 67270 |
Gene Name: | mitochondrial ribosomal protein L42 |
Gene Aliases: |
2700009F22Rik, 2900055D03Rik, D10Ertd322e, HSPC204, L31mt, MRP-L31, PTD007, Rpml31 |
Location: |
Chr.10:95480805-95501992 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl42 | NM_026065.3 | NP_080341.1 | ||
| AK002530.1 | BAB22166.1 | |||
| AK003427.2 | BAB22785.1 | |||
| AK012227.1 | ||||
| AK013693.1 | BAB28957.1 | |||
| BC024337.1 | AAH24337.1 |
More Information
Set Membership: |
|

Genomic Map