Assay Details
Target Gene Details
Entrez Gene ID: | 69090 |
Gene Name: | activating signal cointegrator 1 complex subunit 1 |
Gene Aliases: |
1810015P09Rik, AI550520, ASC1p50, CGI-18 |
Location: |
Chr.10:60002493-60099990 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ascc1 | NM_001199187.2 | NP_001186116.1 | ||
NM_026937.3 | NP_081213.1 | |||
AK007519.1 | BAB25087.1 | |||
AK139944.1 | BAE24191.1 | |||
AK171958.1 | BAE42747.1 | |||
AW457357.1 | ||||
BC030905.1 | AAH30905.1 |
