Assay Details
Target Gene Details
Entrez Gene ID: | 14421 |
Gene Name: | beta-1,4-N-acetyl-galactosaminyl transferase 1 |
Gene Aliases: |
4933429D13Rik, Gal-NAc-T, GalNAc-T, GalNAcT, Galgt1, Ggm-2, Ggm2 |
Location: |
Chr.10:127165156-127172340 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4galnt1 | NM_008080.5 | NP_032106.1 | ||
| NM_027739.2 | NP_082015.2 | |||
| NR_045166.1 | ||||
| AK144484.1 | BAE25913.1 | |||
| BC036996.1 | AAH36996.1 | |||
| BC057199.1 | AAH57199.1 | |||
| L25885.1 | AAA65027.1 | |||
| U18975.1 | AAA85496.1 |

Genomic Map